Methyltriethoxysilane ≥98.0%
Supplier: TCI America
Synonyms:
Triethoxy(methyl)silane
M0451-25ML
M0451-500ML
TCM0451-25MLEA
15.63
USD
TCM0451-25ML
TCM0451-500ML
Methyltriethoxysilane ≥98.0%
Methyltriethoxysilane
CAS Number: 2031-67-6
MDL Number: MFCD00009066
Molecular Formula: C7H18O3Si
Molecular Weight: 178.30
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 143
Flash Point (°C): 23
Specific Gravity (20/20): 0.90
Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Methyltriethoxysilane
Formula:
C₇H₁₈O₃Si MW: 178.3 g/mol Boiling Pt: 143 °C Melting Pt: –50 °C Density: 0.90 (20 °C) Flash Pt: 23 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009066 CAS Number: 2031-67-6 EINECS: 217-983-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...