S-Methylisothiouronium sulfate

Supplier: TCI America

Synonyms: 2-Methyl-2-thiopseudourea sulfateS-Methylisothiourea sulfate2-Methyl-2-thiopseudourea hemisulfateS-Methylisothiourea sulphateS-Methylthiouronium sulfateS-Methylthiouronium sulphate2-Methyl-2-thiopseudourea sulphate2-Methyl-2-thiopseudourea hemisulphate

M0442-25G M0442-500G M0442-100G
TCM0442-025GEA 16.75 USD
TCM0442-025G TCM0442-100G TCM0442-500G
S-Methylisothiouronium sulfate
S-Methylisothiouronium sulphate

CAS Number: 867-44-7
MDL Number: MFCD00013055
Molecular Formula: C2H6N2S
Molecular Weight: 278.36
Form: Crystal
Color: White


Other Product Names / Synonyms: 2-Methyl-2-thiopseudourea Sulfate

Formula: C₄H₁₄N₄O₄S₃
MW: 278.38 g/mol
MDL Number: MFCD00013055
CAS Number: 867-44-7
EINECS: 212-759-7
https://us.vwr.com/stibo/web/std.lang.all/38/81/28973881.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR