(±)-2-Methyltetrahydrofuran ≥98.0% stabilized
Supplier: TCI America
|
Danger
|
Stabilization:
Stabilised with Stabilised
M0437-100ML
M0437-25ML
M0437-500ML
TCM0437-25MLEA
18.98
USD
TCM0437-25ML
TCM0437-500ML
TCM0437-100ML
(±)-2-Methyltetrahydrofuran ≥98.0% stabilized
(±)-2-Methyltetrahydrofuran
(stabilized with BHT)
CAS Number: 96-47-9
MDL Number: MFCD00005367
Molecular Formula: C5H10O
Molecular Weight: 86.13
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 78
Flash Point (°C): -11
Specific Gravity (20/20): 0.86
Other Product Names / Synonyms: Tetrahydro-2-methylfuran
Formula:
C₅H₁₀O MW: 86.13 g/mol Boiling Pt: 78 °C Melting Pt: –136 °C Density: 0.86 (20 °C) Flash Pt: –11 °C |
MDL Number:
MFCD00005367 CAS Number: 96-47-9 EINECS: 202-507-4 UN: 2536 ADR: 3,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...