α-Methylstyrene ≥99.0% stabilized

Supplier: TCI America
Danger

Synonyms: 2-Phenylpropene Isopropenylbenzene

Stabilization: Stabilised with Stabilised

M0429-500ML M0429-25ML
TCM0429-500MLEA 36.72 USD
TCM0429-500ML TCM0429-25ML
α-Methylstyrene ≥99.0% stabilized
α-Methylstyrene

(stabilized with TBC)
CAS Number: 98-83-9
MDL Number: MFCD00008859
Molecular Formula: C9H10
Molecular Weight: 118.18
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 162
Melting point (°C): -23
Flash Point (°C): 48
Specific Gravity (20/20): 0.91
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Isopropenylbenzene, 2-Phenyl-1-propene

Formula: C₉H₁₀
MW: 118.18 g/mol
Boiling Pt: 162 °C
Melting Pt: –23 °C
Density: 0.91 (20 °C)
Flash Pt: 48 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008859
CAS Number: 98-83-9
EINECS: 202-705-0
UN: 2303
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/38/65/28973865.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR