2-Methyl-8-quinolinol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 8-Hydroxyquinaldine

M0420-100G M0420-500G M0420-25G
TCM0420-100GEA 86 USD
TCM0420-100G TCM0420-500G TCM0420-025G
2-Methyl-8-quinolinol ≥98.0% (by GC, titration analysis)
2-Methyl-8-quinolinol

CAS Number: 826-81-3
MDL Number: MFCD00006765
Molecular Formula: C10H9NO
Molecular Weight: 159.19
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 145
Melting point (°C): 73


Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: 8-Hydroxy-2-methylquinoline, 2-Methyloxine, 8-Hydroxyquinaldine

Formula: C₁₀H₉NO
MW: 159.19 g/mol
Boiling Pt: 145 °C
Melting Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00006765
CAS Number: 826-81-3
EINECS: 212-562-6
UN: 3077
ADR: 9,III
https://us.vwr.com/stibo/web/std.lang.all/38/50/28973850.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR