2-Methylnaphthalene ≥96.0%

Supplier: TCI America

Synonyms: β-Methylnaphthalene

M0372-25G M0372-500G
TCM0372-25GEA 17.7 USD
TCM0372-25G TCM0372-500G
2-Methylnaphthalene ≥96.0%
2-Methylnaphthalene

CAS Number: 91-57-6
MDL Number: MFCD00004118
Molecular Formula: C11H10
Molecular Weight: 142.20
Purity/Analysis Method: 96.0% (GC)
Form: Crystal
Boiling point (°C): 241
Flash Point (°C): 97
Freezing point (°C): 31

Formula: C₁₁H₁₀
MW: 142.2 g/mol
Boiling Pt: 241 °C
Melting Pt: 32…35 °C
Density: 1.0058 g/cm³ (20 °C)
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00004118
CAS Number: 91-57-6
EINECS: 202-078-3
UN: 3077
ADR: 9,III
https://us.vwr.com/stibo/web/std.lang.all/37/92/28973792.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR