Citraconic anhydride ≥98.0%

Supplier: TCI America

Synonyms: 3-Methylfuran-2,5-dione

M0365-500G M0365-25G
TCM0365-25GEA 24.35 USD
TCM0365-25G TCM0365-500G
Citraconic anhydride ≥98.0%
Citraconic anhydride

CAS Number: 616-02-4
MDL Number: MFCD00005522
Molecular Formula: C5H4O3
Molecular Weight: 112.08
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 215
Flash Point (°C): 101
Freezing point (°C): 7
Specific Gravity (20/20): 1.25


Product Categories: Bioscience, Functional Group Blocking Reagents
Other Product Names / Synonyms: Methylmaleic Anhydride

Formula: C₅H₄O₃
MW: 112.08 g/mol
Boiling Pt: 215 °C
Melting Pt: 7…8 °C
Density: 1.25 (20 °C)
Flash Pt: 101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005522
CAS Number: 616-02-4
EINECS: 210-459-0
UN: 2810
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/37/82/28973782.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR