3-Methylglutaric acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America

M0330-5G M0330-25G
TCM0330-005GEA 84.25 USD
TCM0330-005G TCM0330-25G
3-Methylglutaric acid ≥99.0% (by titrimetric analysis)
3-Methylglutaric acid

CAS Number: 626-51-7
MDL Number: MFCD00002727
Molecular Formula: C6H10O4
Molecular Weight: 146.14
Purity/Analysis Method: 99.0% (T)
Form: Crystal
Melting point (°C): 87


Other Product Names / Synonyms: 3-Methylpentanedioic Acid

Formula: C₆H₁₀O₄
MW: 146.1412 g/mol
Melting Pt: 87 °C
MDL Number: MFCD00002727
CAS Number: 626-51-7
EINECS: 210-951-5
https://us.vwr.com/stibo/web/std.lang.all/37/35/28973735.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR