1-Methylisoquinoline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

M0298-100MG M0298-1G
TCM0298-100MGEA 20.3 USD
TCM0298-100MG TCM0298-001G
1-Methylisoquinoline ≥98.0% (by GC, titration analysis)
1-Methylisoquinoline

CAS Number: 1721-93-3
MDL Number: MFCD00006902
Molecular Formula: C10H9N
Molecular Weight: 143.19
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 255
Flash Point (°C): 102
Specific Gravity (20/20): 1.08

Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 255 °C
Melting Pt: 10 °C
Density: 1.08 (20 °C)
Flash Pt: 102 °C
MDL Number: MFCD00006902
CAS Number: 1721-93-3
EINECS: 217-017-6
https://us.vwr.com/stibo/web/std.lang.all/37/01/28973701.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR