5-Methyl-2-octanone ≥80.0%

Supplier: TCI America

M0278-500ML M0278-25ML
TCM0278-500MLEA 790 USD
TCM0278-500ML TCM0278-025ML
5-Methyl-2-octanone ≥80.0%
5-Methyl-2-octanone

CAS Number: 58654-67-4
MDL Number: MFCD00059414
Molecular Formula: C9H18O
Molecular Weight: 142.24
Purity/Analysis Method: 80.0% (GC)
Form: Clear Liquid
Boiling point (°C): 102
Specific Gravity (20/20): 0.83


Other Product Names / Synonyms: Methyl 3-Methylhexyl Ketone

Formula: C₉H₁₈O
MW: 142.24 g/mol
Boiling Pt: 102 °C
Density: 0.83 (20 °C)
MDL Number: MFCD00059414
CAS Number: 58654-67-4
EINECS: 261-382-4
https://us.vwr.com/stibo/web/std.lang.all/36/86/28973686.jpg

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR