Methylcyclohexane ≥99.0%
Supplier: TCI America
|
Danger
|
M0244-100ML
TCM0244-100MLEA
66.07
USD
TCM0244-100ML
Methylcyclohexane ≥99.0%
Methylcyclohexane
[for Spectrophotometry]
CAS Number: 108-87-2
MDL Number: MFCD00001497
Molecular Formula: C7H14
Molecular Weight: 98.19
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 100
Melting point (°C): -127
Flash Point (°C): -4
Specific Gravity (20/20): 0.77
Product Categories: Analytical Science, Solvents for Spectrophotometry
Other Product Names / Synonyms: Hexahydrotoluene, MCH
Formula:
C₆H₁₁CH₃ MW: 98.19 g/mol Boiling Pt: 100 °C Melting Pt: –127 °C Density: 0.77 (20 °C) Flash Pt: –4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001497 CAS Number: 108-87-2 EINECS: 203-624-3 UN: 2296 ADR: 3,II Merck Index: 12,10412 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...