3-Methylanisole ≥98.0%

Supplier: TCI America

Synonyms: 1-Methoxy-3-methylbenzene

M0151-500ML M0151-25ML
TCM0151-25MLEA 23.89 USD
TCM0151-25ML TCM0151-500ML
3-Methylanisole ≥98.0%
3-Methylanisole

CAS Number: 100-84-5
MDL Number: MFCD00008395
Molecular Formula: C8H10O
Molecular Weight: 122.17
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 177
Flash Point (°C): 54
Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: m-Cresol Methyl Ether, 3-Methylanisole

Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 177 °C
Melting Pt: –47 °C
Density: 0.97 (20 °C)
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00008395
CAS Number: 100-84-5
EINECS: 202-893-4
UN: 3271
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/34/51/28973451.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR