(±)-3-Methoxy-1,2-propanediol ≥98.0%

Supplier: TCI America

Synonyms: (±)-3-Methoxy-1,2-dihydroxy propane

M0125-25ML
TCM0125-025MLEA 139.26 USD
TCM0125-025ML
(±)-3-Methoxy-1,2-propanediol ≥98.0%
(±)-3-Methoxy-1,2-propanediol

CAS Number: 623-39-2
MDL Number: MFCD00004719
Molecular Formula: C4H10O3
Molecular Weight: 106.12
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 220
Flash Point (°C): 113
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: Glycerol alpha-Monomethyl Ether

Formula: C₄H₁₀O₃
MW: 106.12 g/mol
Boiling Pt: 220 °C
Density: 1.12 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00004719
CAS Number: 623-39-2
EINECS: 210-791-6
https://us.vwr.com/stibo/web/std.lang.all/34/14/28973414.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR