2-Dimethylaminoethyl methacrylate ≥98.5% stabilized
Supplier: TCI America
Synonyms:
DMA
, N,N-Dimethylaminoethyl methacrylate
Stabilization:
Stabilised with Stabilised
(stabilized with MEHQ)
CAS Number: 2867-47-2
MDL Number: MFCD00008589
Molecular Formula: C8H15NO2
Molecular Weight: 157.21
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 70
Melting point (°C): -30
Flash Point (°C): 57
Specific Gravity (20/20): 0.93
Product Categories: Materials Chemistry, Monomers, Bioscience, Monomers for Synthetic Polymers (for DDS)
Other Product Names / Synonyms: Methacrylic Acid 2-(Dimethylamino)ethyl Ester
Formula:
C₈H₁₅NO₂ MW: 157.21 g/mol Boiling Pt: 70 °C Melting Pt: –30 °C Density: 0.93 (20 °C) Flash Pt: 57 °C |
MDL Number:
MFCD00008589 CAS Number: 2867-47-2 EINECS: 220-688-8 UN: 2522 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...