Manganese (II) acetylacetonate dihydrate ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Manganese(II) Acetylacetonate
, Bis(2,4-pentanedionato)manganese(II) dihydrate
, Acetylacetone manganese(II) salt
M0042-25G
TCM0042-025GEA
101.46
USD
TCM0042-025G
Manganese (II) acetylacetonate dihydrate ≥97.0% (by titrimetric analysis)
Manganese (II) acetylacetonate dihydrate
CAS Number: 14024-58-9
MDL Number: MFCD00000022
Molecular Formula: C10H14MnO4
Molecular Weight: 253.16
Purity/Analysis Method:
Form: Crystal
Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Oxidation, Materials Chemistry, Magnetic Metal Complexes (Magnetic Materials)
Other Product Names / Synonyms: Acetylacetone Manganese(II) Salt, Manganese(II) Acetylacetonate
Formula:
C₁₀H₁₄MnO₄·2H₂O |
MDL Number:
MFCD00000022 CAS Number: 14024-58-9 EINECS: 237-858-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...