Dimethyl maleate ≥95.0%

Supplier: TCI America
Warning

Synonyms: Maleic acid dimethyl ester

M0012-500ML M0012-25ML
TCM0012-500MLEA 61.89 USD
TCM0012-500ML TCM0012-25ML
Dimethyl maleate ≥95.0%
Dimethyl maleate

CAS Number: 624-48-6
MDL Number: MFCD00008459
Molecular Formula: C6H8O4
Molecular Weight: 144.13
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 205
Melting point (°C): -19
Flash Point (°C): 103
Specific Gravity (20/20): 1.15


Product Categories: Materials Chemistry, Resin Additives, Copolymerization Monomers
Other Product Names / Synonyms: Maleic Acid Dimethyl Ester, Methyl Maleate

Formula: C₆H₈O₄
MW: 144.13 g/mol
Boiling Pt: 205 °C
Melting Pt: –19 °C
Density: 1.15 (20 °C)
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00008459
CAS Number: 624-48-6
EINECS: 210-848-5
https://us.vwr.com/stibo/web/std.lang.all/32/72/28973272.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR