L-(-)-tert-Leucine (H-Tle-OH) ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
(S)-2-Amino-3,3-dimethylbutanoic acid
, H-Tle-OH
L0147-25G
L0147-5G
L0147-1G
TCL0147-025GEA
171.54
USD
TCL0147-025G
TCL0147-1G
TCL0147-005G
L-(-)-tert-Leucine (H-Tle-OH) ≥98.0% (by titrimetric analysis)
L(-)-tert-Leucine (H-Tle-OH)
CAS Number: 20859-02-3
MDL Number: MFCD00064218
Molecular Formula: C6H13NO2
Molecular Weight: 131.18
Purity/Analysis Method:
Form: Crystal
Specific rotation [a]20/D: -9 deg (C=3, H2O)
Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (S)-2-Amino-3,3-dimethylbutyric Acid, H-Tle-OH
Formula:
C₆H₁₃NO₂ MW: 131.2 g/mol |
MDL Number:
MFCD00064218 CAS Number: 20859-02-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...