3,6-Dimethyl-1,4-dioxane-2,5-dione ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
DL-Lactide
L0091-25G
L0091-100G
TCL0091-025GEA
119.1
USD
TCL0091-025G
TCL0091-100G
3,6-Dimethyl-1,4-dioxane-2,5-dione ≥98.0% (by titrimetric analysis)
3,6-Dimethyl-1,4-dioxane-2,5-dione
CAS Number: 95-96-5
MDL Number: MFCD00011685
Molecular Formula: C6H8O4
Molecular Weight: 144.13
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 142
Melting point (°C): 124
Flash Point (°C): 200
Product Categories: Materials Chemistry, Monomers, Bioscience, Monomers for Synthetic Polymers (for DDS)
Other Product Names / Synonyms: DL-Dilactide, 3,6-Dimethyl-1,4-dioxane-2,5-dione
Formula:
C₆H₈O₄ MW: 144.13 g/mol Boiling Pt: 142 °C Melting Pt: 124 °C Flash Pt: 200 °C |
MDL Number:
MFCD00011685 CAS Number: 95-96-5 EINECS: 202-468-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...