2,4-Dihydroxypteridine (lumazine) ≥99.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Lumazine
, 2,4-Dihydroxypteridine
, 2,4-Pteridinediol
L0086-5G
L0086-1G
TCL0086-5GEA
381.45
USD
TCL0086-5G
TCL0086-001G
2,4-Dihydroxypteridine (lumazine) ≥99.0% (by titrimetric analysis)
2,4-Dihydroxypteridine (Lumazine)
CAS Number: 487-21-8
MDL Number: MFCD00066250
Molecular Formula: C6H4N4O2
Molecular Weight: 164.12
Purity/Analysis Method:
Form: Crystal
Other Product Names / Synonyms: 2,4-Dihydroxypteridine, 2,4-Pteridinediol
Formula:
C₆H₄N₄O₂ MW: 164.12 |
MDL Number:
MFCD00066250 CAS Number: 487-21-8 EINECS: 207-652-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...