3,5-Lutidine ≥98.0%

Supplier: TCI America
Danger

Synonyms: 3,5-Dimethylpyridine

L0068-500ML L0068-25ML
TCL0068-25MLEA 20.1 USD
TCL0068-25ML TCL0068-500ML
3,5-Lutidine ≥98.0%
3,5-Lutidine

CAS Number: 591-22-0
MDL Number: MFCD00006404
Molecular Formula: C7H9N
Molecular Weight: 107.16
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 172
Melting point (°C): -9
Flash Point (°C): 47
Specific Gravity (20/20): 0.94


Product Categories: Synthetic Organic Chemistry, Ligands, Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: 3,5-Dimethylpyridine

Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 172 °C
Melting Pt: –9 °C
Density: 0.94 (20 °C)
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00006404
CAS Number: 591-22-0
EINECS: 209-708-6
UN: 1992
ADR: 3,III

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR