(S)-(-)-4-Isopropyl-2-oxazolidinone ≥98.0%
Supplier: TCI America
Synonyms:
(4S)-2-Oxo-4-(prop-2-yl)-1,3-oxazolidine
, (4S)-(-)-4-Isopropyl-2-oxazolidinone
, (S)-4-Isopropyl-2-oxazolidinone
, (4S)-(-)-Isopropyl-2-oxazolidinone
, (4S)-4-(Prop-2-yl)-1,3-oxazolidin-2-one
I0451-25G
I0451-1G
I0451-5G
TCI0451-005GEA
46.69
USD
TCI0451-005G
TCI0451-001G
TCI0451-25G
(S)-(-)-4-Isopropyl-2-oxazolidinone ≥98.0%
(S)-(-)-4-Isopropyl-2-oxazolidinone
CAS Number: 17016-83-0
MDL Number: MFCD00010847
Molecular Formula: C6H11NO2
Molecular Weight: 129.16
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 72
Specific rotation [a]20/D: -17.5 deg (C=6, EtOH)
Formula:
C₆H₁₁NO₂ MW: 129.16 g/mol Melting Pt: 72 °C |
MDL Number:
MFCD00010847 CAS Number: 17016-83-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...