Benzyl isobutyrate ≥98.0%

Supplier: TCI America

I0424-25ML
TCI0424-025MLEA 19.29 USD
TCI0424-025ML
Benzyl isobutyrate ≥98.0%
Benzyl isobutyrate

CAS Number: 103-28-6
MDL Number: MFCD00048315
Molecular Formula: C11H14O2
Molecular Weight: 178.23
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 230
Flash Point (°C): 100
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: Isobutyric Acid Benzyl Ester

Formula: C₁₁H₁₄O₂
Boiling Pt: 230 °C
Density: 1.01 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00048315
CAS Number: 103-28-6
EINECS: 203-095-9
https://us.vwr.com/stibo/web/std.lang.all/22/65/28972265.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR