N-Isopropylaniline ≥99.0% (by GC, titration analysis)
Supplier: TCI America
I0384-100ML
I0384-25ML
I0384-500ML
TCI0384-025MLEA
58.86
USD
TCI0384-025ML
TCI0384-500ML
TCI0384-100ML
N-Isopropylaniline ≥99.0% (by GC, titration analysis)
N-Isopropylaniline
CAS Number: 768-52-5
MDL Number: MFCD00026347
Molecular Formula: C9H13N
Molecular Weight: 135.21
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 213
Melting point (°C): -50
Flash Point (°C): 88
Specific Gravity (20/20): 0.94
Formula:
C₆H₅NHCH(CH₃)₂ MW: 135.21 g/mol Boiling Pt: 213 °C Melting Pt: –50 °C Density: 0.94 (20 °C) Flash Pt: 88 °C |
MDL Number:
MFCD00026347 CAS Number: 768-52-5 EINECS: 212-196-7 UN: 2810 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...