2-Iodoanisole ≥98.0% stabilized

Supplier: TCI America

Synonyms: 1-Iodo-2-methoxybenzene

Stabilization: Stabilised with Stabilised

I0382-25G
TCI0382-025GEA 62 USD
TCI0382-025G
2-Iodoanisole ≥98.0% stabilized
2-Iodoanisole

(stabilized with Copper chip)
CAS Number: 529-28-2
MDL Number: MFCD00001039
Molecular Formula: C7H7IO
Molecular Weight: 234.04
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 238
Flash Point (°C): 110
Specific Gravity (20/20): 1.80

Formula: C₇H₇IO
MW: 234.04 g/mol
Boiling Pt: 238 °C
Density: 1.80 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001039
CAS Number: 529-28-2
EINECS: 208-456-4
Merck Index: 13,05050
https://us.vwr.com/stibo/web/std.lang.all/22/12/28972212.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR