Indene ≥98.0%

Supplier: TCI America
Danger

Synonyms: 1H-Indene

I0354-25ML
TCI0354-025MLEA 124.85 USD
TCI0354-025ML
Indene ≥98.0%
Indene

CAS Number: 95-13-6
MDL Number: MFCD00003777
Molecular Formula: C9H8
Molecular Weight: 116.16
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 180
Flash Point (°C): 58
Freezing point (°C): -2
Specific Gravity (20/20): 1.00
Storage Temperature: 0°C

Formula: C₉H₈
MW: 116.16 g/mol
Boiling Pt: 180 °C
Melting Pt: –2 °C
Density: 1.00 (20 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00003777
CAS Number: 95-13-6
EINECS: 202-393-6
UN: 3295
ADR: 3,III
Merck Index: 13,04964
https://us.vwr.com/stibo/web/std.lang.all/21/81/28972181.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR