2-Isopropylnaphthalene ≥95.0%
Supplier: TCI America
I0327-25ML
I0327-500ML
TCI0327-025MLEA
61.89
USD
TCI0327-025ML
TCI0327-500ML
2-Isopropylnaphthalene ≥95.0%
2-Isopropylnaphthalene
CAS Number: 2027-17-0
MDL Number: MFCD00014322
Molecular Formula: C13H14
Molecular Weight: 170.26
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 268
Flash Point (°C): 122
Specific Gravity (20/20): 0.98
Formula:
C₁₃H₁₄ MW: 170.25 g/mol Boiling Pt: 268 °C Melting Pt: 11 °C Density: 0.98 (20 °C) Flash Pt: 122 °C |
MDL Number:
MFCD00014322 CAS Number: 2027-17-0 EINECS: 217-976-0 UN: 3077 ADR: 9,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...