1-Iodonaphthalene ≥97.0%

Supplier: TCI America
Warning

I0266-25G
TCI0266-025GEA 89.32 USD
TCI0266-025G
1-Iodonaphthalene ≥97.0%
1-Iodonaphthalene

CAS Number: 90-14-2
MDL Number: MFCD00003876
Molecular Formula: C10H7I
Molecular Weight: 254.07
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 305
Flash Point (°C): 113
Freezing point (°C): 9
Specific Gravity (20/20): 1.74


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 1-Naphthyl Iodide

Formula: C₁₀H₇I
MW: 254.07 g/mol
Boiling Pt: 305 °C
Melting Pt: 7 °C
Density: 1.74 (20 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00003876
CAS Number: 90-14-2
EINECS: 201-968-9
https://us.vwr.com/stibo/web/std.lang.all/20/77/28972077.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR