4-Isopropylphenol ≥99.0%
Supplier: TCI America
|
Danger
|
I0262-25G
I0262-100G
I0262-500G
TCI0262-025GEA
77.14
USD
TCI0262-025G
TCI0262-100G
TCI0262-500G
4-Isopropylphenol ≥99.0%
4-Isopropylphenol
CAS Number: 99-89-8
MDL Number: MFCD00002372
Molecular Formula: C9H12O
Molecular Weight: 136.19
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 109
Melting point (°C): 61
Flash Point (°C): 106
Other Product Names / Synonyms: p-Cumenol, 4-Hydroxycumene
Formula:
C₉H₁₂O MW: 136.19 g/mol Boiling Pt: 109 °C Melting Pt: 61 °C Density: 0.99 g/cm³ (20 °C) Flash Pt: 106 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002372 CAS Number: 99-89-8 EINECS: 202-798-8 UN: 2430 ADR: 8,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...