1-Butyl isothiocyanate ≥98.0%
Supplier: TCI America
Synonyms:
Butyl isothiocyanate
I0249-250ML
I0249-25ML
TCI0249-25MLEA
92.37
USD
TCI0249-25ML
TCI0249-250ML
1-Butyl isothiocyanate ≥98.0%
1-Butyl isothiocyanate
CAS Number: 592-82-5
MDL Number: MFCD00004824
Molecular Formula: C5H9NS
Molecular Weight: 115.19
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 175
Flash Point (°C): 66
Specific Gravity (20/20): 0.96
Other Product Names / Synonyms: Isothiocyanic Acid Butyl Ester
Formula:
C₅H₉NS MW: 115.2 g/mol Boiling Pt: 175 °C Density: 0.96 (20 °C) Flash Pt: 66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004824 CAS Number: 592-82-5 EINECS: 209-770-4 UN: 1693 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...