Ethyl isothiocyanate ≥97.0%
Supplier: TCI America
I0188-25ML
TCI0188-25MLEA
52.33
USD
TCI0188-25ML
Ethyl isothiocyanate ≥97.0%
Ethyl isothiocyanate
CAS Number: 542-85-8
MDL Number: MFCD00004820
Molecular Formula: C3H5NS
Molecular Weight: 87.14
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 132
Flash Point (°C): 24
Specific Gravity (20/20): 1.00
Other Product Names / Synonyms: Isothiocyanic Acid Ethyl Ester
Formula:
C₃H₅NS MW: 87.15 g/mol Boiling Pt: 132 °C Melting Pt: –6 °C Density: 1.00 (20 °C) Flash Pt: 24 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004820 CAS Number: 542-85-8 EINECS: 208-831-2 UN: 2929 ADR: 6.1,II Merck Index: 13,03848 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...