Isobutyl isothiocyanate ≥97.0%

Supplier: TCI America
Danger

Synonyms: 2-Methylpropyl isothiocyanate

I0186-25ML
TCI0186-025MLEA 78 USD
TCI0186-025ML
Isobutyl isothiocyanate ≥97.0%
Isobutyl isothiocyanate

CAS Number: 591-82-2
MDL Number: MFCD00022062
Molecular Formula: C5H9NS
Molecular Weight: 115.19
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 164
Flash Point (°C): 24
Specific Gravity (20/20): 0.95


Other Product Names / Synonyms: Isothiocyanic Acid Isobutyl Ester

Formula: C₅H₉NS
MW: 115.2 g/mol
Boiling Pt: 164 °C
Density: 0.95 (20 °C)
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00022062
CAS Number: 591-82-2
EINECS: 209-733-2
UN: 2924
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/19/78/28971978.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR