2-Isopropylphenol ≥98.0%

Supplier: TCI America
Danger

I0173-25G I0173-500G
TCI0173-025GEA 22.66 USD
TCI0173-025G TCI0173-500G
2-Isopropylphenol ≥98.0%
2-Isopropylphenol

CAS Number: 88-69-7
MDL Number: MFCD00002224
Molecular Formula: C9H12O
Molecular Weight: 136.19
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 214
Flash Point (°C): 104
Freezing point (°C): 16
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: o-Cumenol, 2-Hydroxycumene

Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 214 °C
Melting Pt: 14…16 °C
Density: 1.00 (20 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00002224
CAS Number: 88-69-7
EINECS: 201-852-8
UN: 3145
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/19/63/28971963.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR