4-Isopropylbenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

I0169-25G
TCI0169-025GEA 146.26 USD
TCI0169-025G
4-Isopropylbenzoic acid ≥98.0% (by GC, titration analysis)
4-Isopropylbenzoic acid

CAS Number: 536-66-3
MDL Number: MFCD00002564
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 118


Other Product Names / Synonyms: Cuminic Acid

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 118 °C
Density: 1.162 g/cm³ (20 °C)
MDL Number: MFCD00002564
CAS Number: 536-66-3
EINECS: 208-642-5
Merck Index: 12,02684
https://us.vwr.com/stibo/web/std.lang.all/19/60/28971960.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR