Methyl isobutyrate ≥99.0%

Supplier: TCI America

I0106-500ML I0106-25ML
TCI0106-025MLEA 20.84 USD
TCI0106-025ML TCI0106-500ML
Methyl isobutyrate ≥99.0%
Methyl isobutyrate

CAS Number: 547-63-7
MDL Number: MFCD00008914
Molecular Formula: C5H10O2
Molecular Weight: 102.13
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 92
Melting point (°C): -85
Flash Point (°C): 13
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: Isobutyric Acid Methyl Ester

Formula: C₅H₁₀O₂
Boiling Pt: 92 °C
Melting Pt: –85 °C
Density: 0.89 (20 °C)
Flash Pt: 13 °C
MDL Number: MFCD00008914
CAS Number: 547-63-7
EINECS: 208-929-5
UN: 1237
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/18/90/28971890.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR