3,4-Dichlorophenyl isocyanate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
I0092-500G
I0092-25G
TCI0092-500GEA
630.36
USD
TCI0092-500G
TCI0092-25G
3,4-Dichlorophenyl isocyanate ≥98.0% (by GC, titration analysis)
3,4-Dichlorophenyl isocyanate
CAS Number: 102-36-3
MDL Number: MFCD00002017
Molecular Formula: C7H3Cl2NO
Molecular Weight: 188.01
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 120
Melting point (°C): 40
Flash Point (°C): 123
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Isocyanic Acid 3,4-Dichlorophenyl Ester
Formula:
C₇H₃Cl₂NO MW: 188.01 g/mol Boiling Pt: 120 °C Melting Pt: 40 °C Flash Pt: 123 °C |
MDL Number:
MFCD00002017 CAS Number: 102-36-3 EINECS: 203-026-2 UN: 2250 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...