1-Iodo-3-nitrobenzene ≥98.0%

Supplier: TCI America

I0063-25G
TCI0063-025GEA 106.58 USD
TCI0063-025G
1-Iodo-3-nitrobenzene ≥98.0%
1-Iodo-3-nitrobenzene

CAS Number: 645-00-1
MDL Number: MFCD00007218
Molecular Formula: C6H4INO2
Molecular Weight: 249.01
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Flash Point (°C): 72
Freezing point (°C): 33


Other Product Names / Synonyms: 3-Iodonitrobenzene

Formula: C₆H₄INO₂
MW: 249.01 g/mol
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 38 °C
Density: 2.227 g/cm³ (20 °C)
Flash Pt: 72 °C
MDL Number: MFCD00007218
CAS Number: 645-00-1
EINECS: 211-427-9
UN: 2926
ADR: 4.1,III
https://us.vwr.com/stibo/web/std.lang.all/18/49/28971849.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR