Indoline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2,3-Dihydroindol
I0033-250ML
I0033-25ML
TCI0033-025MLEA
42.73
USD
TCI0033-025ML
TCI0033-250ML
Indoline ≥98.0% (by GC, titration analysis)
Indoline
CAS Number: 496-15-1
MDL Number: MFCD00005705
Molecular Formula: C8H9N
Molecular Weight: 119.17
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 231
Flash Point (°C): 93
Specific Gravity (20/20): 1.07
Other Product Names / Synonyms: 2,3-Dihydroindole
Formula:
C₈H₉N MW: 119.17 g/mol Boiling Pt: 231 °C Melting Pt: –21 °C Density: 1.07 (20 °C) Flash Pt: 93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005705 CAS Number: 496-15-1 EINECS: 207-816-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...