3-(Indol-3-yl)propionic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Indole-3-propionic acid

I0032-5G I0032-25G
TCI0032-005GEA 29.36 USD
TCI0032-005G TCI0032-25G
3-(Indol-3-yl)propionic acid ≥98.0% (by titrimetric analysis)
3-(Indol-3-yl)propionic acid

CAS Number: 830-96-6
MDL Number: MFCD00005660
Molecular Formula: C11H11NO2
Molecular Weight: 189.21
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 132


Product Categories: Bioscience, Auxins
Other Product Names / Synonyms: IPA

Formula: C₁₁H₁₁NO₂
MW: 189.21 g/mol
Melting Pt: 132 °C
Density: 1.356 g/cm³ (20 °C)
MDL Number: MFCD00005660
CAS Number: 830-96-6
EINECS: 212-600-1
https://us.vwr.com/stibo/web/std.lang.all/18/20/28971820.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR