Indol-3-ylacetonitrile ≥98.0%

Supplier: TCI America

Synonyms: 3-Indolylacetonitrile3-Indoleacetonitrile2-(1H-Indol-3-yl)acetonitrileIndole-3-Acetonitrile

I0024-25G I0024-1G
TCI0024-001GEA 46.69 USD
TCI0024-001G TCI0024-025G
Indol-3-ylacetonitrile ≥98.0%
Indol-3-ylacetonitrile

CAS Number: 771-51-7
MDL Number: MFCD00005628
Molecular Formula: C10H8N2
Molecular Weight: 156.19
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 37
Flash Point (°C): 113
Storage Temperature: 0-10°C


Product Categories: Bioscience, Auxins
Other Product Names / Synonyms: IAN

Formula: C₁₀H₈N₂
MW: 156.19 g/mol
Melting Pt: 37 °C
Flash Pt: 113 °C
MDL Number: MFCD00005628
CAS Number: 771-51-7
EINECS: 212-232-1
UN: 3439
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/18/12/28971812.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR