Indium(III) acetylacetonate ≥99.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Tris(2,4-pentanedionato-O,O')indium (III)
, Indium (III) tris(2,4-pentanedionato-O,O')
, Indium (III) 2,4-pentanedionate
CAS Number: 14405-45-9
MDL Number: MFCD00013494
Molecular Formula: C15H21InO6
Molecular Weight: 412.15
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 186
Flash Point (°C): 180
Other Product Names / Synonyms: Acetylacetone Indium(III) Salt, Indium(III) Acetylacetonate
Formula:
C₁₅H₂₁InO₆ MW: 412.15 g/mol Melting Pt: 186 °C Flash Pt: 180 °C |
MDL Number:
MFCD00013494 CAS Number: 14405-45-9 EINECS: 238-378-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...