Indane ≥95.0%

Supplier: TCI America

Synonyms: 2,3-Dihydro-1H-indeneIndan

I0011-25ML I0011-100ML I0011-500ML
TCI0011-025MLEA 26.39 USD
TCI0011-025ML TCI0011-500ML TCI0011-100ML
Indane ≥95.0%
Indane

CAS Number: 496-11-7
MDL Number: MFCD00003795
Molecular Formula: C9H10
Molecular Weight: 118.18
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 179
Flash Point (°C): 50
Specific Gravity (20/20): 0.96


Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: 2,3-Dihydro-1H-indene, Hydrindene

Formula: C₉H₁₀
MW: 118.18 g/mol
Boiling Pt: 179 °C
Melting Pt: –51.4 °C
Density: 0.96 (20 °C)
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00003795
CAS Number: 496-11-7
EINECS: 207-814-7
UN: 3295
ADR: 3,III
Merck Index: 13,04956
https://us.vwr.com/stibo/web/std.lang.all/18/00/28971800.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR