1-Bromoperfluorooctane ≥98.0%

Supplier: TCI America
Warning

Synonyms: Perfluorooctyl bromide1-BromoheptadecafluorooctanePerflubron

H0946-5G H0946-25G
TCH0946-025GEA 216.13 USD
TCH0946-025G TCH0946-005G
1-Bromoperfluorooctane ≥98.0%
1-Bromoperfluorooctane

CAS Number: 423-55-2
MDL Number: MFCD00042082
Molecular Formula: C8BrF17
Molecular Weight: 498.96
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 141
Specific Gravity (20/20): 1.93


Product Categories: Synthetic Organic Chemistry, Fluorous Chemistry, Fluorous Compounds, Fluorinated Building Blocks, Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Perfluoro-n-octyl Bromide

Formula: BrF₂C(CF₂)₆CF₃
MW: 498.96 g/mol
Boiling Pt: 141 °C
Melting Pt: 6 °C
Density: 1.93 (20 °C)
MDL Number: MFCD00042082
CAS Number: 423-55-2
EINECS: 207-028-4
Merck Index: 12,07299
https://us.vwr.com/stibo/web/std.lang.all/09/67/28970967.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR