4-Hydrazinobenzoic acid ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
4-Hydrazinylbenzoic acid
H0800-25G
TCH0800-025GEA
49.13
USD
TCH0800-025G
4-Hydrazinobenzoic acid ≥97.0% (by HPLC, titration analysis)
4-Hydrazinobenzoic acid
CAS Number: 619-67-0
MDL Number: MFCD00007581
Molecular Formula: C7H8N2O2
Molecular Weight: 152.15
Purity/Analysis Method:
Form: Crystal
Formula:
C₇H₈N₂O₂ MW: 152.15 g/mol |
MDL Number:
MFCD00007581 CAS Number: 619-67-0 EINECS: 210-609-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...