4-Heptylbenzoic acid

Supplier: TCI America

H0774-10G
TCH0774-010GEA 357 USD
TCH0774-010G
4-Heptylbenzoic acid
4-Heptylbenzoic acid

CAS Number: 38350-87-7
MDL Number: MFCD00009722
Molecular Formula: C14H20O2
Molecular Weight: 220.31
Form: Crystal
Color: White
Melting point (°C): 102

Formula: C₁₄H₂₀O₂
MW: 220.31 g/mol
Boiling Pt: 350 °C (1013 hPa)
Melting Pt: 102 °C
Density: ∼1.114 g/cm³ (20 °C)
MDL Number: MFCD00009722
CAS Number: 38350-87-7
EINECS: 253-894-1
https://us.vwr.com/stibo/web/std.lang.all/08/06/28970806.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR