(±)-δ-Hexalactone ≥99.0%
Supplier: TCI America
Synonyms:
(±)-δ-Caprolactone
, (±)-Tetrahydro-6-methyl-2H-pyran-2-one
, (±)-Hexan-5-olide
, 6-Methyltetrahydro-2H-pyran-2-one
CAS Number: 823-22-3
MDL Number: MFCD00083574
Molecular Formula: C6H10O2
Molecular Weight: 114.14
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 115
Flash Point (°C): 103
Specific Gravity (20/20): 1.05
Product Categories: Materials Chemistry, Monomers, Bioscience, Lipids
Other Product Names / Synonyms: delta-Hexalactone, 5-Methyl-delta-valerolactone, Tetrahydro-6-methyl-2H-pyran-2-one
Formula:
C₆H₁₀O₂ MW: 114.14 g/mol Boiling Pt: 115 °C Density: 1.05 (20 °C) Flash Pt: 103 °C |
MDL Number:
MFCD00083574 CAS Number: 823-22-3 EINECS: 212-511-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...