Diethyl hexafluoroglutarate ≥98.0%
Supplier: TCI America
Synonyms:
Diethyl perfluoroglutarate
CAS Number: 424-40-8
MDL Number: MFCD00015156
Molecular Formula: C9H10F6O4
Molecular Weight: 296.17
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 84
Flash Point (°C): 87
Specific Gravity (20/20): 1.36
Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: Diethyl Hexafluoroglutarate, 2,2,3,3,4,4-Hexafluoropentanedioic Acid Diethyl Ester, Hexafluoroglutaric Acid Diethyl Ester
Formula:
C₉H₁₀F₆O₄ MW: 296.17 g/mol Boiling Pt: 84 °C Density: 1.36 (20 °C) Flash Pt: 87 °C |
MDL Number:
MFCD00015156 CAS Number: 424-40-8 EINECS: 207-035-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...