3-Piperidinemethanol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-(Hydroxymethyl)piperidine

H0737-25G
TCH0737-025GEA 57 USD
TCH0737-025G
3-Piperidinemethanol ≥98.0% (by GC, titration analysis)
3-Piperidinemethanol

CAS Number: 4606-65-9
MDL Number: MFCD00005997
Molecular Formula: C6H13NO
Molecular Weight: 115.18
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 107
Melting point (°C): 60
Flash Point (°C): 131


Other Product Names / Synonyms: 3-(Hydroxymethyl)piperidine

Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 107 °C
Melting Pt: 60 °C
Flash Pt: 131 °C
MDL Number: MFCD00005997
CAS Number: 4606-65-9
EINECS: 225-013-0
https://us.vwr.com/stibo/web/std.lang.all/07/74/28970774.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR