MOPSO (3-(4-morpholino)-2-hydroxypropanesulfonic acid) ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 2-Hydroxy-3-morpholinopropane-1-sulphonic acid2-Hydroxy-3-morpholinopropane-1-sulfonic acid

H0671-25G
TCH0671-025GEA 34.56 USD
TCH0671-025G
MOPSO (3-(4-morpholino)-2-hydroxypropanesulfonic acid) ≥98.0% (by titrimetric analysis)
MOPSO (3-(4-Morpholino)-2-hydroxypropanesulphonic acid)

CAS Number: 68399-77-9
MDL Number: MFCD00009629
Molecular Formula: C7H15NO5S
Molecular Weight: 225.26
Purity/Analysis Method: 98.0% (T)
Form: Crystal


Product Categories: Bioscience, Good's Buffering Agents
Other Product Names / Synonyms: 3-(N-Morpholino)-2-hydroxypropanesulfonic Acid, MOPSO

Formula: C₇H₁₅NO₅S
MW: 225.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009629
CAS Number: 68399-77-9
EINECS: 269-989-6
https://us.vwr.com/stibo/web/std.lang.all/06/89/28970689.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR