(4-Hydroxyphenyl)acetonitrile ≥99.0%
Supplier: TCI America
Synonyms:
4-Hydroxybenzyl cyanide
, 2-(4-Hydroxyphenyl)acetonitrile
H0657-500G
H0657-25G
TCH0657-500GEA
452.17
USD
TCH0657-500G
TCH0657-025G
(4-Hydroxyphenyl)acetonitrile ≥99.0%
(4-Hydroxyphenyl)acetonitrile
CAS Number: 14191-95-8
MDL Number: MFCD00002383
Molecular Formula: C8H7NO
Molecular Weight: 133.15
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 150
Melting point (°C): 71
Other Product Names / Synonyms: 4-Hydroxyphenylacetonitrile
Formula:
C₈H₇NO MW: 133.15 g/mol Boiling Pt: 150 °C Melting Pt: 71 °C |
MDL Number:
MFCD00002383 CAS Number: 14191-95-8 EINECS: 238-046-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...