EPPS (HEPPS, 4-(2-Hydroxyethyl)-1-piperazinepropanesulfonic acid) ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: HEPPS4-(2-Hydroxyethyl)-1-piperazinepropanesulphonic acid3-[4-(2-Hydroxyethyl)-1-piperazinyl]propanesulfonic acid4-(2-Hydroxyethyl)piperazine-1-propanesulphonic acid3-[4-(2-Hydroxyethyl)-1-piperazinyl]propanesulphonic acid4-(2-Hydroxyethyl)piperazine-1-propanesulfonic acidN-(2-Hydroxyethyl)piperazine-N′-(3-propanesulfonic acid)N-(2-Hydroxyethyl)piperazine-N′-(3-propanesulphonic acid)3-[4-(2-Hydroxyethyl)-1-piperazinyl]-propanesulfonic acid4-(2-Hydroxyethyl)-1-piperazinepropanesulfonic acid3-[4-(2-Hydroxyethyl)-1-piperazinyl]-propanesulphonic acid

H0576-25G
TCH0576-025GEA 118.44 USD
TCH0576-025G
EPPS (HEPPS, 4-(2-Hydroxyethyl)-1-piperazinepropanesulfonic acid) ≥98.0% (by titrimetric analysis)
EPPS (HEPPS, 4-(2-Hydroxyethyl)-1-piperazinepropanesulphonic acid)

[Good's buffer component for biological research]
CAS Number: 16052-06-5
MDL Number: MFCD00006160
Molecular Formula: C9H20N2O4S
Molecular Weight: 252.33
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 239


Product Categories: Bioscience, Good's Buffering Agents
Other Product Names / Synonyms: HEPPS, 3-[4-(2-Hydroxyethyl)-1-piperazinyl]propanesulfonic Acid

Formula: C₉H₂₀N₂O₄S
MW: 252.33 g/mol
Melting Pt: 239 °C
Storage Temperature: Ambient
MDL Number: MFCD00006160
CAS Number: 16052-06-5
EINECS: 240-198-8
https://us.vwr.com/stibo/web/std.lang.all/05/91/28970591.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR